Assay Details
| Gene Symbol: NUPR2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001145712.1 | NP_001139184.1 | 1 - 2 | 378 | 107 | |
| GenBank mRNA | BC043523.1 | - | 1 - 2 | 378 | 107 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC07003358.hg.1
Set Membership: |
|
Gene Ontology Categories:
Function(s)
Process(es)
negative regulation of transcription from RNA polymerase II promoter
transcription, DNA-templated
cellular response to DNA damage stimulus
cell cycle arrest
negative regulation of cell proliferation
cellular response to starvation
negative regulation of cell cycle arrest
chromatin binding
negative regulation of transcription from RNA polymerase II promoter
transcription, DNA-templated
cellular response to DNA damage stimulus
cell cycle arrest
negative regulation of cell proliferation
cellular response to starvation
negative regulation of cell cycle arrest
chromatin binding
MicroArray Probes:
Agilent
Affymetrix
ArrayStar
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P25730006 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P05505839 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P25730014 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P25729994 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P25730023 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P05505842 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P25730024 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P05505836 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P25730017 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P05505840 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P05505835 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P25729987 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P05505841 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P25729997 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P25730034 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P05505843 | - | - |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 151268 | AK025460.1 | genbank_mrna_human |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 151268 | BC000971.1 | genbank_mrna_human |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 151268 | BC001618.1 | genbank_mrna_human |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 151268 | BC004863.2 | genbank_mrna_human |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 151268 | BC016645.2 | genbank_mrna_human |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 151268 | BC018129.1 | genbank_mrna_human |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 151268 | CR595610.1 | genbank_mrna_human |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 151268 | CR619649.1 | genbank_mrna_human |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 151268 | NM_021154.3 | refseq_mrna_human_NM |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 151268 | NM_058179.2 | refseq_mrna_human_NM |
| Agilent Whole Human Genome Microarray 4x44K v2 | A_32_P42964 | - | - |
| Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_HS_44k_2078 | - | - |
| Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_HS_44k_2077 | - | - |
| Agilent Whole Human Genome Oligo Microarray 4x44K | 35183 | - | - |
| Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 12035 | - | - |
| Agilent Whole Human Genome Microarray 4x44K v2 | A_32_P42964 | - | - |
| SurePrint G3 Human GE Microarray G4851A | A_32_P42964 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25729988 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25729993 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25730002 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25730006 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25729997 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25729977 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05505832 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25730024 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P15945320 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05505842 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05505841 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05505834 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05505833 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25730023 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25729987 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05505840 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05505839 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25730017 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25729994 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05505836 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05505843 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25730014 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05505835 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05505841 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25729994 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25729997 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25730002 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05505839 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05505835 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25729977 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25730023 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05505836 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05505834 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25729988 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05505832 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05505842 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25730017 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25729987 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05505833 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25730006 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05505843 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25730014 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25730024 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P15945320 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25729993 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05505840 | - | - |
| Agilent Human Genome CGH Microarray 4x44K | A_14_P201856 | - | - |
| Agilent Human 1B Microarray G4111A | A_32_P174220 | - | - |
| Agilent Human 1B Microarray G4111A | A_32_P42971 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P05505843 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P15945297 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P15945291 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P05505822 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P05505845 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P15945288 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P05505827 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P05505847 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P05505832 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P05505840 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P05505815 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P05505835 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P05505836 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P15945307 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P15945294 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P05505842 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P15945289 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P05505828 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P05505818 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P15945305 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P05505841 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P15945303 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P05505829 | - | - |
| Agilent SurePrint G3 Human CGH Microarray 2x400K | A_16_P38036691 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Gene 1.0 ST Array | 8139764 | - | - |
| Affymetrix Human Gene 1.1 ST Array | 8139763 | - | - |
| Affymetrix Human Genome U219 Array | 11737153_at | - | - |
| Affymetrix Human Genome U219 Array | 11737153_PM_at | - | - |
| Affymetrix Human Transcriptome Array 2.0 | PSR07021848.hg.1 | TC07003358.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR07021848.hg.1 | ENST00000329309.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR07021848.hg.1 | NM_001145712.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR07021849.hg.1 | TC07003358.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR07021849.hg.1 | ENST00000329309.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR07021849.hg.1 | NM_001145712.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011167.hg.1 | ENST00000329309.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011167.hg.1 | ENST00000329309.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011167.hg.1 | ENST00000329309.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011167.hg.1 | NM_001145712.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011167.hg.1 | NM_001145712.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011167.hg.1 | NM_001145712.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700179840.hg.1 | ENST00000329309.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700179840.hg.1 | NM_001145712.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700179840.hg.1 | ENST00000329309.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700179840.hg.1 | NM_001145712.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700179840.hg.1 | TC0700011167.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0700011167.hg.1 | ENST00000329309.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700011167.hg.1 | ENST00000329309.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700011167.hg.1 | ENST00000329309.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700011167.hg.1 | NM_001145712.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700011167.hg.1 | NM_001145712.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700011167.hg.1 | NM_001145712.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700179838.hg.1 | ENST00000329309.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700179838.hg.1 | NM_001145712.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700179838.hg.1 | ENST00000329309.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700179838.hg.1 | NM_001145712.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700179838.hg.1 | TC0700011167.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0700179841.hg.1 | ENST00000329309.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700179841.hg.1 | NM_001145712.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700179841.hg.1 | ENST00000329309.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700179841.hg.1 | NM_001145712.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700179841.hg.1 | TC0700011167.hg.1 | Clariom D TC |
| Human Genome U95 Set | 69239_PM_f_at | - | - |
| Human Genome U95 Set | 69239_f_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A032840 | - | - |
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A032840 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| HumanHT-12 v4 Expression BeadChip | ILMN_3295235 | NM_001145712.1 | human_transcriptome |
| HumanHT-12 v4 Expression BeadChip | ILMN_3295235 | BC043523.1 | human_transcriptome |

NUPR2
Genomic Map