Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR12015284.hg.1 |
ENST00000519357.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12015284.hg.1 |
ENST00000523102.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12015284.hg.1 |
ENST00000524270.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12015284.hg.1 |
NM_001146316.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR12015284.hg.1 |
NM_032641.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR12015284.hg.1 |
TC12001152.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012742.hg.1 |
ENST00000519357.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012742.hg.1 |
ENST00000519357.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012742.hg.1 |
ENST00000519357.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012742.hg.1 |
ENST00000519357.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012742.hg.1 |
ENST00000519357.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012742.hg.1 |
ENST00000523102.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012742.hg.1 |
ENST00000523102.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012742.hg.1 |
ENST00000523102.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012742.hg.1 |
ENST00000523102.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012742.hg.1 |
ENST00000523102.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012742.hg.1 |
ENST00000523102.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012742.hg.1 |
ENST00000524270.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012742.hg.1 |
ENST00000524270.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012742.hg.1 |
ENST00000524270.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012742.hg.1 |
ENST00000524270.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012742.hg.1 |
ENST00000524270.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012742.hg.1 |
ENST00000524270.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012742.hg.1 |
NM_001146316.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012742.hg.1 |
NM_001146316.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012742.hg.1 |
NM_001146316.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012742.hg.1 |
NM_001146316.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012742.hg.1 |
NM_001146316.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012742.hg.1 |
NM_001146316.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012742.hg.1 |
NM_032641.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012742.hg.1 |
NM_032641.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012742.hg.1 |
NM_032641.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012742.hg.1 |
NM_032641.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012742.hg.1 |
NM_032641.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012742.hg.1 |
NM_032641.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR12015286.hg.1 |
ENST00000519357.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12015286.hg.1 |
ENST00000524270.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12015286.hg.1 |
NM_032641.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR12015286.hg.1 |
TC12001152.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
TC1200012742.hg.1 |
ENST00000519357.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012742.hg.1 |
ENST00000519357.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012742.hg.1 |
ENST00000519357.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012742.hg.1 |
ENST00000519357.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012742.hg.1 |
ENST00000519357.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012742.hg.1 |
ENST00000523102.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012742.hg.1 |
ENST00000523102.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012742.hg.1 |
ENST00000523102.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012742.hg.1 |
ENST00000523102.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012742.hg.1 |
ENST00000523102.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012742.hg.1 |
ENST00000523102.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012742.hg.1 |
ENST00000524270.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012742.hg.1 |
ENST00000524270.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012742.hg.1 |
ENST00000524270.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012742.hg.1 |
ENST00000524270.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012742.hg.1 |
ENST00000524270.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012742.hg.1 |
ENST00000524270.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012742.hg.1 |
NM_001146316.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012742.hg.1 |
NM_001146316.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012742.hg.1 |
NM_001146316.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012742.hg.1 |
NM_001146316.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012742.hg.1 |
NM_001146316.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012742.hg.1 |
NM_001146316.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012742.hg.1 |
NM_032641.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012742.hg.1 |
NM_032641.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012742.hg.1 |
NM_032641.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012742.hg.1 |
NM_032641.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012742.hg.1 |
NM_032641.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012742.hg.1 |
NM_032641.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200202358.hg.1 |
ENST00000519357.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200202358.hg.1 |
ENST00000524270.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200202358.hg.1 |
NM_032641.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200202358.hg.1 |
ENST00000519357.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200202358.hg.1 |
ENST00000524270.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200202358.hg.1 |
NM_032641.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200202358.hg.1 |
TC1200012742.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1200202356.hg.1 |
ENST00000519357.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200202356.hg.1 |
ENST00000523102.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200202356.hg.1 |
ENST00000524270.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200202356.hg.1 |
NM_001146316.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200202356.hg.1 |
NM_032641.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200202356.hg.1 |
ENST00000519357.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200202356.hg.1 |
ENST00000523102.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200202356.hg.1 |
ENST00000524270.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200202356.hg.1 |
NM_001146316.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200202356.hg.1 |
NM_032641.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200202356.hg.1 |
TC1200012742.hg.1 |
Clariom D TC |