Assay Details
Gene Symbol: PDLIM5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AF116705.2 | - | 1 | 84 | 91 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | hematopoietic prostaglandin D synthase | HPGDS |
GSTS, GSTS1, GSTS1-1, PGD2, PGDS |
-232.1kb | Anti-sense | Chr. 4 - 94298556 - 94342876 on NCBI Build GRCh38 |
Downstream | BMPR1B antisense RNA 1 (head to head) | BMPR1B-AS1 | 167.3kb | Anti-sense | Chr. 4 - 94743668 - 94757533 on NCBI Build GRCh38 |

More Information
Additional Information:
This assay is in the 5'UTR region of the AF116705.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
