Assay Details
| Gene Symbol: GPSM2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001321039.1 | NP_001307968.1 | 16 | 4853 | 96 | |
| GenBank mRNA | AB445462.1 | - | 16 | 4855 | 96 | |
| AK000053.1 | - | 1 | 1053 | 96 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | chloride channel CLIC like 1 | CLCC1 | MCLC | Anti-sense | Chr. 1 - 108929508 - 108963499 on NCBI Build GRCh38 | |
| Downstream | WD repeat domain 47 | WDR47 | 35.8kb | Anti-sense | Chr. 1 - 108970211 - 109042228 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
|

GPSM2
Genomic Map