Assay Details
Gene Symbol: LOC101929340 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_120577.1 | 1 - 2 | 279 | 96 | ||
GenBank mRNA | AK000493.1 | - | 1 - 2 | 285 | 96 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | endothelial cell adhesion molecule | ESAM | W117m | Anti-sense | Chr. 11 - 124753123 - 124762327 on NCBI Build GRCh38 | |
Downstream | Myb/SANT DNA binding domain containing 2 | MSANTD2 | C11orf61 | 556.0 | Anti-sense | Chr. 11 - 124766492 - 124800879 on NCBI Build GRCh38 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
