Assay Details
| Gene Symbol: NPAS3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK002000.1 | - | 1 | 557 | 105 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | A-kinase anchoring protein 6 | AKAP6 |
ADAP100, ADAP6, AKAP100, PRKA6, mAKAP |
-528.9kb | Sense | Chr. 14 - 32329273 - 32833999 on NCBI Build GRCh38 |
| Downstream | egl-9 family hypoxia inducible factor 3 | EGLN3 |
HIFP4H3, HIFPH3, PHD3 |
559.0kb | Anti-sense | Chr. 14 - 33924215 - 33951078 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

NPAS3
Genomic Map