Assay Details
Gene Symbol: MTMR14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK021608.1 | - | 1 | 1649 | 73 |
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Transcript Location | Amplicon Length | |
---|---|---|---|---|---|---|---|
GenBankmRNA | AU144768.1 | - | 1 | Chr. 3 - 9698320 - 9698868 [-] on Build GRCh38 | 73 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | lipoma HMGIC fusion partner-like 4 | LHFPL4 | -143.1kb | Anti-sense | Chr. 3 - 9498361 - 9553802 on NCBI Build GRCh38 | |
Downstream | copine family member 9 | CPNE9 | 4.9kb | Sense | Chr. 3 - 9703807 - 9729908 on NCBI Build GRCh38 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() |
