Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010563.hg.1 |
ENST00000562742.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010563.hg.1 |
ENST00000562742.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010563.hg.1 |
ENST00000562742.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010563.hg.1 |
ENST00000563754.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010563.hg.1 |
ENST00000563754.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010563.hg.1 |
ENST00000563754.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010563.hg.1 |
ENST00000563754.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010563.hg.1 |
ENST00000563754.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010563.hg.1 |
ENST00000563754.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010563.hg.1 |
NR_110918.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010563.hg.1 |
NR_110918.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010563.hg.1 |
NR_110918.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010563.hg.1 |
NR_110918.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010563.hg.1 |
NR_110918.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010563.hg.1 |
NR_110918.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600181157.hg.1 |
ENST00000563754.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600181157.hg.1 |
NR_110918.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600181157.hg.1 |
ENST00000563754.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600181157.hg.1 |
NR_110918.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600181157.hg.1 |
TC1600010563.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1600010563.hg.1 |
ENST00000562742.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600010563.hg.1 |
ENST00000562742.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600010563.hg.1 |
ENST00000562742.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600010563.hg.1 |
ENST00000563754.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600010563.hg.1 |
ENST00000563754.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600010563.hg.1 |
ENST00000563754.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600010563.hg.1 |
ENST00000563754.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600010563.hg.1 |
ENST00000563754.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600010563.hg.1 |
ENST00000563754.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600010563.hg.1 |
NR_110918.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600010563.hg.1 |
NR_110918.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600010563.hg.1 |
NR_110918.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600010563.hg.1 |
NR_110918.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600010563.hg.1 |
NR_110918.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600010563.hg.1 |
NR_110918.1 |
Refseq |