Assay Details
| Gene Symbol: RIF1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_011511395.2 | XP_011509697.1 | 35 | 9976 | 86 | |
| GenBank mRNA | AL117611.1 | - | 1 | 50 | 86 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | TNF alpha induced protein 6 | TNFAIP6 |
TSG-6, TSG6 |
-99.0kb | Sense | Chr. 2 - 151357592 - 151380048 on NCBI Build GRCh38 |
| Downstream | nebulin | NEB |
NEB177D, NEM2 |
3.2kb | Anti-sense | Chr. 2 - 151485334 - 151734487 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

RIF1
Genomic Map