Assay Details
Gene Symbol: LOC105375640 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | U29109.1 | - | 1 | 151 | 119 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | triple QxxK/R motif containing | TRIQK |
C8orf83, PRO0845, UPF0599 |
-18.5kb | Sense | Chr. 8 - 92883530 - 92966150 on NCBI Build GRCh38 |
Downstream | microRNA 8084 | MIR8084 | hsa-mir-8084 | 44.9kb | Anti-sense | Chr. 8 - 93029751 - 93029839 on NCBI Build GRCh38 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
