Assay Details
Gene Symbol: IGHV3-75 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | Y11327.1 | - | 1 | 150 | 106 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | uncharacterized LOC317730 | C14orf99 | -13.5kb | Sense | Chr. 14 - 106809028 - 106810171 on NCBI Build GRCh38 | |
Downstream | microRNA 5195 | MIR5195 | 27.0kb | Sense | Chr. 14 - 106850885 - 106850999 on NCBI Build GRCh38 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
