Assay Details
| Gene Symbol: GABRA2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_011513675.1 | XP_011511977.1 | 11 | 7938 | 116 | |
| XM_011513676.1 | XP_011511978.1 | 11 | 7914 | 116 | ||
| GenBank mRNA | AF086092.1 | - | 1 | 145 | 116 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | DNA polymerase beta | POLB | Sense | Chr. 8 - 42338452 - 42371813 on NCBI Build GRCh38 | ||
| Downstream | cytochrome c oxidase subunit 7B2 | COX7B2 | 490.6kb | Sense | Chr. 4 - 46734827 - 46909266 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

GABRA2
Genomic Map