Assay Details
| Gene Symbol: INTS6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_011535040.2 | XP_011533342.1 | 19 | 3657 | 105 | |
| GenBank mRNA | AF086440.1 | - | 1 | 243 | 105 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | microRNA 5693 | MIR5693 | -5.2kb | Sense | Chr. 13 - 51348567 - 51348639 on NCBI Build GRCh38 | |
| Downstream | INTS6 antisense RNA 1 | INTS6-AS1 | 99.0kb | Anti-sense | Chr. 13 - 51453345 - 51454716 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

INTS6
Genomic Map