| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR05006378.hg.1 |
TC05000451.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR05006378.hg.1 |
ENST00000327111.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR05006378.hg.1 |
ENST00000506162.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR05006378.hg.1 |
NM_005654.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008086.hg.1 |
ENST00000327111.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008086.hg.1 |
ENST00000327111.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008086.hg.1 |
ENST00000327111.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008086.hg.1 |
ENST00000327111.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008086.hg.1 |
ENST00000327111.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008086.hg.1 |
ENST00000327111.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008086.hg.1 |
ENST00000327111.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008086.hg.1 |
ENST00000327111.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008086.hg.1 |
ENST00000327111.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008086.hg.1 |
ENST00000327111.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008086.hg.1 |
ENST00000327111.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008086.hg.1 |
ENST00000327111.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008086.hg.1 |
ENST00000327111.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008086.hg.1 |
ENST00000327111.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008086.hg.1 |
ENST00000502982.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008086.hg.1 |
ENST00000502982.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008086.hg.1 |
ENST00000502982.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008086.hg.1 |
ENST00000506162.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008086.hg.1 |
ENST00000506162.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008086.hg.1 |
ENST00000506162.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008086.hg.1 |
ENST00000506162.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008086.hg.1 |
ENST00000506162.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008086.hg.1 |
ENST00000506162.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008086.hg.1 |
ENST00000512697.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008086.hg.1 |
ENST00000512697.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008086.hg.1 |
ENST00000615873.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008086.hg.1 |
ENST00000615873.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008086.hg.1 |
ENST00000615873.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008086.hg.1 |
ENST00000615873.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008086.hg.1 |
ENST00000615873.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008086.hg.1 |
ENST00000615873.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008086.hg.1 |
ENST00000615873.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008086.hg.1 |
ENST00000615873.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008086.hg.1 |
NM_005654.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008086.hg.1 |
NM_005654.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008086.hg.1 |
NM_005654.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008086.hg.1 |
NM_005654.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008086.hg.1 |
NM_005654.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008086.hg.1 |
NM_005654.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008086.hg.1 |
NM_005654.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008086.hg.1 |
NM_005654.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008086.hg.1 |
NM_005654.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008086.hg.1 |
NM_005654.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008086.hg.1 |
NM_005654.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008086.hg.1 |
NM_005654.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008086.hg.1 |
NM_005654.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008086.hg.1 |
NM_005654.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0500155379.hg.1 |
ENST00000327111.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500155379.hg.1 |
ENST00000506162.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500155379.hg.1 |
NM_005654.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0500155379.hg.1 |
ENST00000327111.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500155379.hg.1 |
ENST00000506162.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500155379.hg.1 |
NM_005654.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0500155379.hg.1 |
TC0500008086.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC0500008086.hg.1 |
ENST00000327111.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500008086.hg.1 |
ENST00000327111.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500008086.hg.1 |
ENST00000327111.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500008086.hg.1 |
ENST00000327111.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500008086.hg.1 |
ENST00000327111.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500008086.hg.1 |
ENST00000327111.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500008086.hg.1 |
ENST00000327111.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500008086.hg.1 |
ENST00000327111.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500008086.hg.1 |
ENST00000327111.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500008086.hg.1 |
ENST00000327111.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500008086.hg.1 |
ENST00000327111.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500008086.hg.1 |
ENST00000327111.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500008086.hg.1 |
ENST00000327111.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500008086.hg.1 |
ENST00000327111.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500008086.hg.1 |
ENST00000502982.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500008086.hg.1 |
ENST00000502982.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500008086.hg.1 |
ENST00000502982.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500008086.hg.1 |
ENST00000506162.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500008086.hg.1 |
ENST00000506162.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500008086.hg.1 |
ENST00000506162.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500008086.hg.1 |
ENST00000506162.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500008086.hg.1 |
ENST00000506162.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500008086.hg.1 |
ENST00000506162.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500008086.hg.1 |
ENST00000512697.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500008086.hg.1 |
ENST00000512697.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500008086.hg.1 |
ENST00000615873.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500008086.hg.1 |
ENST00000615873.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500008086.hg.1 |
ENST00000615873.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500008086.hg.1 |
ENST00000615873.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500008086.hg.1 |
ENST00000615873.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500008086.hg.1 |
ENST00000615873.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500008086.hg.1 |
ENST00000615873.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500008086.hg.1 |
ENST00000615873.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500008086.hg.1 |
NM_005654.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500008086.hg.1 |
NM_005654.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500008086.hg.1 |
NM_005654.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500008086.hg.1 |
NM_005654.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500008086.hg.1 |
NM_005654.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500008086.hg.1 |
NM_005654.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500008086.hg.1 |
NM_005654.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500008086.hg.1 |
NM_005654.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500008086.hg.1 |
NM_005654.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500008086.hg.1 |
NM_005654.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500008086.hg.1 |
NM_005654.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500008086.hg.1 |
NM_005654.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500008086.hg.1 |
NM_005654.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500008086.hg.1 |
NM_005654.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0500155377.hg.1 |
ENST00000327111.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500155377.hg.1 |
ENST00000506162.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500155377.hg.1 |
ENST00000615873.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500155377.hg.1 |
NM_005654.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0500155377.hg.1 |
ENST00000327111.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500155377.hg.1 |
ENST00000506162.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500155377.hg.1 |
ENST00000615873.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500155377.hg.1 |
NM_005654.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0500155377.hg.1 |
TC0500008086.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0500155381.hg.1 |
ENST00000327111.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500155381.hg.1 |
NM_005654.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0500155381.hg.1 |
ENST00000327111.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500155381.hg.1 |
NM_005654.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0500155381.hg.1 |
TC0500008086.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0500155380.hg.1 |
ENST00000327111.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500155380.hg.1 |
ENST00000506162.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500155380.hg.1 |
NM_005654.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0500155380.hg.1 |
ENST00000327111.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500155380.hg.1 |
ENST00000506162.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500155380.hg.1 |
NM_005654.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0500155380.hg.1 |
TC0500008086.hg.1 |
Clariom D TC |