Assay Details
| Gene Symbol: FMNL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AJ008124.1 | - | 1 | 43 | 61 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | uncharacterized LOC339192 | LOC339192 | -25.1kb | Anti-sense | Chr. 17 - 45219313 - 45219974 on NCBI Build GRCh38 | |
| Downstream | spermatogenesis associated 32 | SPATA32 |
AEP2, C17orf46, TEX34, VAD1.2 |
2.1kb | Anti-sense | Chr. 17 - 45254393 - 45262112 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

FMNL1
Genomic Map