Assay Details
Gene Symbol: MAP3K14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AJ008146.1 | - | 1 | 50 | 67 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | MAP3K14 antisense RNA 1 | MAP3K14-AS1 | -5.6kb | Anti-sense | Chr. 17 - 45247925 - 45268630 on NCBI Build GRCh38 | |
Downstream | Rho GTPase activating protein 27 | ARHGAP27 |
CAMGAP1, PP905, SH3D20, SH3P20 |
119.6kb | Sense | Chr. 17 - 45393902 - 45432916 on NCBI Build GRCh38 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() |
