Assay Details
Gene Symbol: ZNF800 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | EF212246.1 | - | 1 | 48 | 98 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | glutamate metabotropic receptor 8 | GRM8 |
GLUR8, GPRC1H, MGLUR8, mGlu8 |
-119.5kb | Sense | Chr. 7 - 126438598 - 127253294 on NCBI Build GRCh38 |
Downstream | uncharacterized LOC100506682 | LOC100506682 | 104.0kb | Anti-sense | Chr. 7 - 127476883 - 127485804 on NCBI Build GRCh38 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() |
