Assay Details
Gene Symbol: SLC37A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AF289589.1 | - | 1 | 750 | 98 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | uncharacterized LOC101928212 | LOC101928212 | -5.3kb | Anti-sense | Chr. 21 - 42569024 - 42573810 on NCBI Build GRCh38 | |
Downstream | uncharacterized LOC101928233 | LOC101928233 | 18.1kb | Anti-sense | Chr. 21 - 42599404 - 42615095 on NCBI Build GRCh38 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() |
