Assay Details
Gene Symbol: PRKCE | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK022272.1 | - | 1 | 1651 | 81 |
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Transcript Location | Amplicon Length | |
---|---|---|---|---|---|---|---|
GenBankmRNA | AU147538.1 | - | 1 | Chr. 2 - 45937160 - 45937718 [-] on Build GRCh38 | 81 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | collagen alpha-2(I) chain-like | LOC102724965 | -284.7kb | Anti-sense | Chr. 2 - 45648733 - 45652047 on NCBI Build GRCh38 | |
Downstream | endothelial PAS domain protein 1 | EPAS1 |
ECYT4, HIF2A, HLF, MOP2, PASD2, bHLHe73 |
359.7kb | Sense | Chr. 2 - 46297402 - 46386703 on NCBI Build GRCh38 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() |
