Assay Details
| Gene Symbol: MCM8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017028105.1 | XP_016883594.1 | 19 | 7466 | 113 | |
| XM_017028107.1 | XP_016883596.1 | 12 | 7556 | 113 | ||
| GenBank mRNA | AK022738.1 | - | 1 | 2251 | 113 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | MCM8 antisense RNA 1 | MCM8-AS1 | Anti-sense | Chr. 20 - 5990795 - 6005821 on NCBI Build GRCh38 | ||
| Downstream | cardiolipin synthase 1 | CRLS1 |
C20orf155, CLS, CLS1, GCD10, dJ967N21.6 |
7.1kb | Sense | Chr. 20 - 6005938 - 6040053 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

MCM8
Genomic Map