Assay Details
| Gene Symbol: RPS6KA6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_014496.4 | NP_055311.1 | 22 | 5794 | 93 | |
| XM_017029423.1 | XP_016884912.1 | 22 | 6964 | 93 | ||
| GenBank mRNA | AK023104.1 | - | 1 | 2056 | 93 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | cylicin 1 | CYLC1 | CYCL1 | -173.6kb | Anti-sense | Chr. X - 83861126 - 83886700 on NCBI Build GRCh38 |
| Downstream | microRNA 548i-4 | MIR548I4 |
MIR548I-4, MIRN548I4, hsa-mir-548i-4 |
162.9kb | Sense | Chr. X - 84225752 - 84225828 on NCBI Build GRCh38 |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
protein phosphorylation
DNA damage response, signal transduction by p53 class mediator resulting in transcription of p21 class mediator
signal transduction
central nervous system development
negative regulation of embryonic development
negative regulation of ERK1 and ERK2 cascade
negative regulation of mesoderm development
magnesium ion binding
protein kinase activity
protein serine/threonine kinase activity
ATP binding
kinase activity
protein phosphorylation
DNA damage response, signal transduction by p53 class mediator resulting in transcription of p21 class mediator
signal transduction
central nervous system development
negative regulation of embryonic development
negative regulation of ERK1 and ERK2 cascade
negative regulation of mesoderm development
magnesium ion binding
protein kinase activity
protein serine/threonine kinase activity
ATP binding
kinase activity
MicroArray Probes:
Affymetrix
ABI
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010196.hg.1 | ENST00000262752.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010196.hg.1 | ENST00000262752.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010196.hg.1 | ENST00000262752.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010196.hg.1 | ENST00000262752.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010196.hg.1 | ENST00000262752.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010196.hg.1 | ENST00000262752.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010196.hg.1 | ENST00000262752.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010196.hg.1 | ENST00000262752.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010196.hg.1 | ENST00000262752.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010196.hg.1 | ENST00000262752.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010196.hg.1 | ENST00000262752.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010196.hg.1 | ENST00000262752.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010196.hg.1 | ENST00000262752.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010196.hg.1 | ENST00000262752.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010196.hg.1 | ENST00000262752.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010196.hg.1 | ENST00000262752.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010196.hg.1 | ENST00000262752.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010196.hg.1 | ENST00000262752.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010196.hg.1 | ENST00000262752.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010196.hg.1 | ENST00000262752.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010196.hg.1 | ENST00000262752.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010196.hg.1 | ENST00000262752.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010196.hg.1 | ENST00000262752.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010196.hg.1 | ENST00000262752.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010196.hg.1 | ENST00000262752.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010196.hg.1 | ENST00000262752.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010196.hg.1 | ENST00000460730.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010196.hg.1 | ENST00000460730.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010196.hg.1 | ENST00000460730.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010196.hg.1 | ENST00000460730.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010196.hg.1 | ENST00000460730.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010196.hg.1 | ENST00000460730.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010196.hg.1 | ENST00000460730.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010196.hg.1 | ENST00000495332.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010196.hg.1 | ENST00000495332.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010196.hg.1 | ENST00000495332.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010196.hg.1 | ENST00000495332.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010196.hg.1 | ENST00000495332.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010196.hg.1 | ENST00000495332.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010196.hg.1 | ENST00000620340.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010196.hg.1 | ENST00000620340.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010196.hg.1 | ENST00000620340.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010196.hg.1 | ENST00000620340.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010196.hg.1 | ENST00000620340.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010196.hg.1 | ENST00000620340.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010196.hg.1 | ENST00000620340.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010196.hg.1 | ENST00000620340.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010196.hg.1 | ENST00000620340.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010196.hg.1 | ENST00000620340.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010196.hg.1 | ENST00000620340.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010196.hg.1 | ENST00000620340.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010196.hg.1 | ENST00000620340.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010196.hg.1 | ENST00000620340.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010196.hg.1 | ENST00000620340.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010196.hg.1 | ENST00000620340.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010196.hg.1 | ENST00000620340.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010196.hg.1 | ENST00000620340.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010196.hg.1 | ENST00000620340.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010196.hg.1 | ENST00000620340.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010196.hg.1 | ENST00000620340.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010196.hg.1 | ENST00000620340.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010196.hg.1 | ENST00000620340.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010196.hg.1 | ENST00000620340.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010196.hg.1 | ENST00000620340.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010196.hg.1 | ENST00000620340.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010196.hg.1 | ENST00000620340.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010196.hg.1 | ENST00000620340.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010196.hg.1 | ENST00000620340.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010196.hg.1 | NM_014496.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010196.hg.1 | NM_014496.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010196.hg.1 | NM_014496.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010196.hg.1 | NM_014496.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010196.hg.1 | NM_014496.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010196.hg.1 | NM_014496.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010196.hg.1 | NM_014496.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010196.hg.1 | NM_014496.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010196.hg.1 | NM_014496.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010196.hg.1 | NM_014496.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010196.hg.1 | NM_014496.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010196.hg.1 | NM_014496.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010196.hg.1 | NM_014496.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010196.hg.1 | NM_014496.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010196.hg.1 | NM_014496.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010196.hg.1 | NM_014496.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010196.hg.1 | NM_014496.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010196.hg.1 | NM_014496.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010196.hg.1 | NM_014496.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010196.hg.1 | NM_014496.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010196.hg.1 | NM_014496.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010196.hg.1 | NM_014496.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010196.hg.1 | NM_014496.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010196.hg.1 | NM_014496.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010196.hg.1 | NM_014496.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010196.hg.1 | NM_014496.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010196.hg.1 | NM_014496.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010196.hg.1 | NM_014496.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010196.hg.1 | NM_014496.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010196.hg.1 | ENST00000262752.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010196.hg.1 | ENST00000262752.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010196.hg.1 | ENST00000262752.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010196.hg.1 | ENST00000262752.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010196.hg.1 | ENST00000262752.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010196.hg.1 | ENST00000262752.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010196.hg.1 | ENST00000262752.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010196.hg.1 | ENST00000262752.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010196.hg.1 | ENST00000262752.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010196.hg.1 | ENST00000262752.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010196.hg.1 | ENST00000262752.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010196.hg.1 | ENST00000262752.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010196.hg.1 | ENST00000262752.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010196.hg.1 | ENST00000262752.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010196.hg.1 | ENST00000262752.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010196.hg.1 | ENST00000262752.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010196.hg.1 | ENST00000262752.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010196.hg.1 | ENST00000262752.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010196.hg.1 | ENST00000262752.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010196.hg.1 | ENST00000262752.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010196.hg.1 | ENST00000262752.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010196.hg.1 | ENST00000262752.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010196.hg.1 | ENST00000262752.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010196.hg.1 | ENST00000262752.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010196.hg.1 | ENST00000262752.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010196.hg.1 | ENST00000262752.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010196.hg.1 | ENST00000460730.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010196.hg.1 | ENST00000460730.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010196.hg.1 | ENST00000460730.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010196.hg.1 | ENST00000460730.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010196.hg.1 | ENST00000460730.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010196.hg.1 | ENST00000460730.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010196.hg.1 | ENST00000460730.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010196.hg.1 | ENST00000495332.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010196.hg.1 | ENST00000495332.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010196.hg.1 | ENST00000495332.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010196.hg.1 | ENST00000495332.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010196.hg.1 | ENST00000495332.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010196.hg.1 | ENST00000495332.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010196.hg.1 | ENST00000620340.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010196.hg.1 | ENST00000620340.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010196.hg.1 | ENST00000620340.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010196.hg.1 | ENST00000620340.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010196.hg.1 | ENST00000620340.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010196.hg.1 | ENST00000620340.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010196.hg.1 | ENST00000620340.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010196.hg.1 | ENST00000620340.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010196.hg.1 | ENST00000620340.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010196.hg.1 | ENST00000620340.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010196.hg.1 | ENST00000620340.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010196.hg.1 | ENST00000620340.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010196.hg.1 | ENST00000620340.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010196.hg.1 | ENST00000620340.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010196.hg.1 | ENST00000620340.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010196.hg.1 | ENST00000620340.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010196.hg.1 | ENST00000620340.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010196.hg.1 | ENST00000620340.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010196.hg.1 | ENST00000620340.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010196.hg.1 | ENST00000620340.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010196.hg.1 | ENST00000620340.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010196.hg.1 | ENST00000620340.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010196.hg.1 | ENST00000620340.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010196.hg.1 | ENST00000620340.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010196.hg.1 | ENST00000620340.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010196.hg.1 | ENST00000620340.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010196.hg.1 | ENST00000620340.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010196.hg.1 | ENST00000620340.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010196.hg.1 | ENST00000620340.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010196.hg.1 | NM_014496.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010196.hg.1 | NM_014496.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010196.hg.1 | NM_014496.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010196.hg.1 | NM_014496.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010196.hg.1 | NM_014496.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010196.hg.1 | NM_014496.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010196.hg.1 | NM_014496.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010196.hg.1 | NM_014496.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010196.hg.1 | NM_014496.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010196.hg.1 | NM_014496.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010196.hg.1 | NM_014496.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010196.hg.1 | NM_014496.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010196.hg.1 | NM_014496.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010196.hg.1 | NM_014496.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010196.hg.1 | NM_014496.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010196.hg.1 | NM_014496.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010196.hg.1 | NM_014496.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010196.hg.1 | NM_014496.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010196.hg.1 | NM_014496.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010196.hg.1 | NM_014496.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010196.hg.1 | NM_014496.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010196.hg.1 | NM_014496.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010196.hg.1 | NM_014496.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010196.hg.1 | NM_014496.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010196.hg.1 | NM_014496.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010196.hg.1 | NM_014496.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010196.hg.1 | NM_014496.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010196.hg.1 | NM_014496.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010196.hg.1 | NM_014496.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00169527.hg.1 | ENST00000620340.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00169527.hg.1 | NM_014496.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00169527.hg.1 | ENST00000620340.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00169527.hg.1 | NM_014496.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00169527.hg.1 | TC0X00010196.hg.1 | Clariom D TC |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 107430 | - | - |

RPS6KA6
Genomic Map