Assay Details
| Gene Symbol: TMEM87A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK023569.1 | - | 1 | 1351 | 72 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
VPS39, HOPS complex subunit |
VPS39 |
TLP, VAM6, hVam6p |
-47.6kb | Sense | Chr. 15 - 42158701 - 42208331 on NCBI Build GRCh38 |
| Downstream |
glucosidase alpha, neutral C |
GANC | 15.6kb | Anti-sense | Chr. 15 - 42273658 - 42353666 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

TMEM87A
Genomic Map