Assay Details
| Gene Symbol: ITGA3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK090532.1 | - | 1 | 54 | 81 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | uncharacterized LOC284080 | LOC284080 | -26.9kb | Anti-sense | Chr. 17 - 50050349 - 50055739 on NCBI Build GRCh38 | |
| Downstream | pyruvate dehydrogenase kinase 2 | PDK2 |
PDHK2, PDKII |
9.8kb | Sense | Chr. 17 - 50094737 - 50111369 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

ITGA3
Genomic Map