Assay Details
| Gene Symbol: ZC3H14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_011537162.2 | XP_011535464.1 | 10 | 7207 | 96 | |
| GenBank mRNA | AL832086.1 | - | 1 | 3048 | 96 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
protein tyrosine phosphatase, non-receptor type 21 |
PTPN21 |
PTPD1, PTPRL10 |
-34.5kb | Anti-sense | Chr. 14 - 88465778 - 88554779 on NCBI Build GRCh38 |
| Downstream | echinoderm microtubule associated protein like 5 | EML5 | EMAP-2 | 20.0kb | Anti-sense | Chr. 14 - 88614213 - 88792752 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

ZC3H14
Genomic Map