Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007683.hg.1 |
ENST00000460946.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007683.hg.1 |
ENST00000460946.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007683.hg.1 |
ENST00000476308.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007683.hg.1 |
ENST00000476308.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007683.hg.1 |
ENST00000482609.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007683.hg.1 |
ENST00000482609.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007683.hg.1 |
ENST00000482609.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007683.hg.1 |
ENST00000482609.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007683.hg.1 |
ENST00000482609.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007683.hg.1 |
ENST00000482609.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007683.hg.1 |
ENST00000482609.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007683.hg.1 |
ENST00000482609.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007683.hg.1 |
ENST00000482609.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007683.hg.1 |
ENST00000482609.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007683.hg.1 |
NR_045697.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007683.hg.1 |
NR_045697.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007683.hg.1 |
NR_045697.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007683.hg.1 |
NR_045697.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007683.hg.1 |
NR_045697.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007683.hg.1 |
NR_045697.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007683.hg.1 |
NR_045697.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007683.hg.1 |
NR_045697.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007683.hg.1 |
NR_045697.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007683.hg.1 |
NR_045697.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR03008521.hg.1 |
TC03000396.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR03008521.hg.1 |
ENST00000482609.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03008521.hg.1 |
NR_045697.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR03008522.hg.1 |
TC03000396.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR03008522.hg.1 |
ENST00000482609.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03008522.hg.1 |
NR_045697.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300155553.hg.1 |
ENST00000482609.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300155553.hg.1 |
NR_045697.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300155553.hg.1 |
ENST00000482609.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300155553.hg.1 |
NR_045697.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300155553.hg.1 |
TC0300007683.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0300007683.hg.1 |
ENST00000460946.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007683.hg.1 |
ENST00000460946.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007683.hg.1 |
ENST00000476308.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007683.hg.1 |
ENST00000476308.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007683.hg.1 |
ENST00000482609.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007683.hg.1 |
ENST00000482609.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007683.hg.1 |
ENST00000482609.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007683.hg.1 |
ENST00000482609.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007683.hg.1 |
ENST00000482609.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007683.hg.1 |
ENST00000482609.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007683.hg.1 |
ENST00000482609.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007683.hg.1 |
ENST00000482609.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007683.hg.1 |
ENST00000482609.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007683.hg.1 |
ENST00000482609.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007683.hg.1 |
NR_045697.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300007683.hg.1 |
NR_045697.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300007683.hg.1 |
NR_045697.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300007683.hg.1 |
NR_045697.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300007683.hg.1 |
NR_045697.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300007683.hg.1 |
NR_045697.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300007683.hg.1 |
NR_045697.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300007683.hg.1 |
NR_045697.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300007683.hg.1 |
NR_045697.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300007683.hg.1 |
NR_045697.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300155551.hg.1 |
ENST00000482609.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300155551.hg.1 |
NR_045697.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300155551.hg.1 |
ENST00000482609.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300155551.hg.1 |
NR_045697.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300155551.hg.1 |
TC0300007683.hg.1 |
Clariom D TC |