Assay Details
Gene Symbol: PRICKLE2-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_045697.1 | 2 - 3 | 296 | 103 | ||
GenBank mRNA | AL832189.1 | - | 2 - 3 | 296 | 103 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | long intergenic non-protein coding RNA 994 | LINC00994 | -7.7kb | Anti-sense | Chr. 3 - 64078361 - 64087363 on NCBI Build GRCh38 | |
Downstream | PRICKLE2 antisense RNA 2 | PRICKLE2-AS2 | 335.0 | Sense | Chr. 3 - 64103470 - 64106358 on NCBI Build GRCh38 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC03000396.hg.1
Set Membership: |
![]() ![]() ![]() |
MicroArray Probes:
Agilent
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Human Whole Genome ChIP Tiling Array | A_17_P22431678 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P02296738 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P22431694 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P22431656 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P02296731 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P22431693 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P22431643 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P22431644 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P22431624 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P22431634 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P22431613 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P22431662 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P02296702 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P02296742 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P22431607 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P15446089 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P02296668 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P22431615 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P02296672 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P22431650 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P02296712 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P15446090 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P02296710 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P22431661 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P22431683 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P22431666 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P02296714 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P02296669 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P02296698 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P22431682 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P02296648 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P15446088 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P22431626 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P02296708 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P22431648 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P02296744 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P22431691 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P22431609 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P02296706 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P22431647 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P02296746 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P22431640 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P02296661 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P22431688 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P02296713 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P15446083 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P02296679 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P02296691 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P22431687 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P22431679 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P02296688 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P02296666 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P02296682 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P22431703 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P22431612 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P02296658 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P22431639 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P22431675 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P02296655 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P22431653 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P22431677 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P22431633 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P02296675 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P22431646 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P02296740 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P22431621 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P22431663 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P02296722 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P22431618 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P02296678 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P02296732 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P22431645 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P22431674 | - | - |
Agilent SurePrint G3 Human CGH Microarray 4x180K | 132860 | - | - |
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 544054 | - | - |
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 879253 | - | - |
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 518897 | - | - |
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 841592 | - | - |
Agilent Human Genome CGH + SNP Microarray | A_16_P16254634 | - | - |
Agilent Human Genome CGH Microarray 105A (G4412A) | A_16_P00722717 | - | - |
Agilent Human Genome 244K CGH Microarray (Alpha Test) | A_16_P16254634 | - | - |
Agilent Human Genome 244K CGH Microarray (Alpha Test) | A_16_P00722733 | - | - |
Agilent Human Genome 244K CGH Microarray (Alpha Test) | A_16_P00722717 | - | - |
Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 | 160992 | - | - |
Agilent HD CGH Microarray (2X105K) Oxford design AMADID#019015 | 14617 | - | - |
Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P00722725 | - | - |
Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P00722717 | - | - |
Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P16254634 | - | - |
Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P00722706 | - | - |
Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P00722707 | - | - |
Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P00722721 | - | - |
Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P16254631 | - | - |
Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P00722740 | - | - |
Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P16254686 | - | - |
Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P36297142 | - | - |
Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P36297093 | - | - |
Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P16254648 | - | - |
Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P00722733 | - | - |
Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P31032564 | - | - |
Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P00722738 | - | - |
Agilent SurePrint G3 Human CGH Microarray 2x400K | A_16_P16254634 | - | - |
Agilent SurePrint G3 Human CGH Microarray 2x400K | A_16_P00722740 | - | - |
Agilent SurePrint G3 Human CGH Microarray 2x400K | A_16_P36297093 | - | - |
Agilent SurePrint G3 Human CGH Microarray 2x400K | A_16_P00722733 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR03008523.hg.1 | TC03000396.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR03008523.hg.1 | ENST00000482609.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR03008523.hg.1 | NR_045697.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0300155555.hg.1 | ENST00000482609.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300155555.hg.1 | NR_045697.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0300155555.hg.1 | ENST00000482609.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300155555.hg.1 | NR_045697.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0300155555.hg.1 | TC0300007683.hg.1 | Clariom D TC |
