Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008621.hg.1 |
ENST00000567359.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008621.hg.1 |
ENST00000567359.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008621.hg.1 |
ENST00000567359.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008621.hg.1 |
NR_110938.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008621.hg.1 |
NR_110938.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008621.hg.1 |
NR_110938.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600164333.hg.1 |
ENST00000567359.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600164333.hg.1 |
NR_110938.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600164333.hg.1 |
ENST00000567359.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600164333.hg.1 |
NR_110938.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600164333.hg.1 |
TC1600008621.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1600164334.hg.1 |
ENST00000567359.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600164334.hg.1 |
NR_110938.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600164334.hg.1 |
ENST00000567359.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600164334.hg.1 |
NR_110938.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600164334.hg.1 |
TC1600008621.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1600008621.hg.1 |
ENST00000567359.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008621.hg.1 |
ENST00000567359.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008621.hg.1 |
ENST00000567359.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008621.hg.1 |
NR_110938.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600008621.hg.1 |
NR_110938.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600008621.hg.1 |
NR_110938.1 |
Refseq |