Assay Details
| Gene Symbol: LOC284240 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BC017478.1 | - | 1 | 1248 | 100 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | nuclear factor of activated T-cells 1 | NFATC1 |
NF-ATC, NF-ATc1.2, NFAT2, NFATc |
-47.1kb | Sense | Chr. 18 - 79395772 - 79529323 on NCBI Build GRCh38 |
| Downstream | uncharacterized LOC284241 | LOC284241 | 61.7kb | Anti-sense | Chr. 18 - 79638928 - 79679745 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
|

LOC284240
Genomic Map