Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007591.hg.1 |
ENST00000423681.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007591.hg.1 |
ENST00000423681.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007591.hg.1 |
ENST00000423681.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007591.hg.1 |
NR_121182.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007591.hg.1 |
NR_121182.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007591.hg.1 |
NR_121182.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900151344.hg.1 |
ENST00000423681.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900151344.hg.1 |
NR_121182.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900151344.hg.1 |
ENST00000423681.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900151344.hg.1 |
NR_121182.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900151344.hg.1 |
TC0900007591.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0900007591.hg.1 |
ENST00000423681.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007591.hg.1 |
ENST00000423681.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007591.hg.1 |
ENST00000423681.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007591.hg.1 |
NR_121182.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900007591.hg.1 |
NR_121182.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900007591.hg.1 |
NR_121182.1 |
Refseq |