Assay Details
Gene Symbol: LOC105370613 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | BC032017.1 | - | 1 | 1947 | 66 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | long intergenic non-protein coding RNA 1146 | LINC01146 | -70.9kb | Anti-sense | Chr. 14 - 88024550 - 88087346 on NCBI Build GRCh38 | |
Downstream | uncharacterized LOC105370612 | LOC105370612 | 46.0 | Anti-sense | Chr. 14 - 88160637 - 88180198 on NCBI Build GRCh38 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
