Assay Details
Gene Symbol: MAPK10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001318067.1 | NP_001304996.1 | 13 | 6314 | 65 | |
NM_001318068.1 | NP_001304997.1 | 13 | 6409 | 65 | ||
NM_001318069.1 | NP_001304998.1 | 14 | 6879 | 65 | ||
NM_002753.4 | NP_002744.1 | 14 | 6420 | 65 | ||
NM_138980.3 | NP_620446.1 | 14 | 6906 | 65 | ||
NM_138982.3 | NP_620448.1 | 14 | 6879 | 65 | ||
XM_005263131.2 | XP_005263188.1 | 14 | 6773 | 65 | ||
GenBank mRNA | AF052141.1 | - | 1 | 1445 | 65 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | Rho GTPase activating protein 24 | ARHGAP24 |
FILGAP, RC-GAP72, RCGAP72, p73, p73RhoGAP |
-9.6kb | Anti-sense | Chr. 4 - 85475131 - 86002670 on NCBI Build GRCh38 |
Downstream | uncharacterized LOC101929064 | LOC101929064 | 105.9kb | Anti-sense | Chr. 4 - 86119806 - 86219926 on NCBI Build GRCh38 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
