Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012246.hg.1 |
ENST00000565283.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012246.hg.1 |
ENST00000565283.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012246.hg.1 |
ENST00000565283.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012246.hg.1 |
NR_037631.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012246.hg.1 |
NR_037631.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012246.hg.1 |
NR_037631.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012246.hg.1 |
NR_037631.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200012246.hg.1 |
ENST00000565283.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012246.hg.1 |
ENST00000565283.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012246.hg.1 |
ENST00000565283.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012246.hg.1 |
NR_037631.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200012246.hg.1 |
NR_037631.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200012246.hg.1 |
NR_037631.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200012246.hg.1 |
NR_037631.1 |
Refseq |