Assay Details
| Gene Symbol: POLK | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AF315601.1 | - | 1 | 1250 | 113 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | collagen type IV alpha 3 binding protein | COL4A3BP |
CERT, CERTL, GPBP, MRD34, STARD11 |
-57.0 | Anti-sense | Chr. 5 - 75368486 - 75511981 on NCBI Build GRCh38 |
| Downstream | ankyrin repeat and death domain containing 1B | ANKDD1B | 97.9kb | Sense | Chr. 5 - 75611289 - 75671846 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

POLK
Genomic Map