Assay Details
| Gene Symbol: LOC100287525 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK098252.1 | - | 1 | 1549 | 93 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | gap junction protein delta 3 | GJD3 |
CX31.9, Cx30.2, GJA11, GJC1 |
Anti-sense | Chr. 17 - 40360652 - 40364693 on NCBI Build GRCh38 | |
| Downstream | topoisomerase (DNA) II alpha | TOP2A |
TOP2, TP2A |
23.1kb | Anti-sense | Chr. 17 - 40388521 - 40417950 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

LOC100287525
Genomic Map