Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011121.hg.1 |
ENST00000356215.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011121.hg.1 |
ENST00000356215.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011121.hg.1 |
ENST00000356215.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011121.hg.1 |
ENST00000439236.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011121.hg.1 |
ENST00000439236.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011121.hg.1 |
ENST00000439236.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011121.hg.1 |
ENST00000439236.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011121.hg.1 |
ENST00000504038.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011121.hg.1 |
ENST00000504038.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011121.hg.1 |
ENST00000504038.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011121.hg.1 |
ENST00000536648.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011121.hg.1 |
ENST00000536648.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011121.hg.1 |
ENST00000536648.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011121.hg.1 |
NM_001292023.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011121.hg.1 |
NM_001292023.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011121.hg.1 |
NM_001292023.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011121.hg.1 |
NM_001292023.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011121.hg.1 |
NM_001292024.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011121.hg.1 |
NM_001292024.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011121.hg.1 |
NM_001292024.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011121.hg.1 |
NM_001292024.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011121.hg.1 |
NM_001292024.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011121.hg.1 |
NR_120478.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011121.hg.1 |
NR_120478.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011121.hg.1 |
NR_120478.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200186379.hg.1 |
ENST00000504038.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200186379.hg.1 |
ENST00000536648.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200186379.hg.1 |
NR_120478.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200186379.hg.1 |
ENST00000504038.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200186379.hg.1 |
ENST00000536648.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200186379.hg.1 |
NR_120478.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200186379.hg.1 |
TC1200011121.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1200011121.hg.1 |
ENST00000356215.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011121.hg.1 |
ENST00000356215.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011121.hg.1 |
ENST00000356215.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011121.hg.1 |
ENST00000439236.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011121.hg.1 |
ENST00000439236.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011121.hg.1 |
ENST00000439236.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011121.hg.1 |
ENST00000439236.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011121.hg.1 |
ENST00000504038.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011121.hg.1 |
ENST00000504038.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011121.hg.1 |
ENST00000504038.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011121.hg.1 |
ENST00000536648.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011121.hg.1 |
ENST00000536648.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011121.hg.1 |
ENST00000536648.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011121.hg.1 |
NM_001292023.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011121.hg.1 |
NM_001292023.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011121.hg.1 |
NM_001292023.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011121.hg.1 |
NM_001292023.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011121.hg.1 |
NM_001292024.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011121.hg.1 |
NM_001292024.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011121.hg.1 |
NM_001292024.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011121.hg.1 |
NM_001292024.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011121.hg.1 |
NM_001292024.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011121.hg.1 |
NR_120478.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011121.hg.1 |
NR_120478.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011121.hg.1 |
NR_120478.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200186378.hg.1 |
ENST00000504038.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200186378.hg.1 |
ENST00000536648.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200186378.hg.1 |
NR_120478.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200186378.hg.1 |
ENST00000504038.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200186378.hg.1 |
ENST00000536648.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200186378.hg.1 |
NR_120478.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200186378.hg.1 |
TC1200011121.hg.1 |
Clariom D TC |