Assay Details
| Gene Symbol: CEP152 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BC029603.1 | - | 1 | 1146 | 76 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | fibrillin 1 | FBN1 |
ACMICD, ECTOL1, FBN, GPHYSD2, MASS, MFLS, MFS1, OCTD, SGS, SSKS, WMS, WMS2 |
-164.9kb | Anti-sense | Chr. 15 - 48408306 - 48645788 on NCBI Build GRCh38 |
| Downstream | SHC adaptor protein 4 | SHC4 |
RaLP, SHCD |
11.8kb | Anti-sense | Chr. 15 - 48823737 - 48963444 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

CEP152
Genomic Map