Assay Details
| Gene Symbol: KCTD16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005268492.2 | XP_005268549.1 | 3 | 12640 | 78 | |
| XM_005268493.2 | XP_005268550.1 | 3 | 12036 | 78 | ||
| XM_006714789.2 | XP_006714852.1 | 3 | 12700 | 78 | ||
| XM_011537671.2 | XP_011535973.1 | 5 | 12535 | 78 | ||
| GenBank mRNA | BC036491.1 | - | 1 | 1746 | 78 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | Yip1 domain family member 5 | YIPF5 |
FinGER5, SB140, SMAP-5, SMAP5, YIP1A |
-312.0kb | Anti-sense | Chr. 5 - 144158159 - 144170714 on NCBI Build GRCh38 |
| Downstream | PRELI domain containing 2 | PRELID2 | 852.2kb | Anti-sense | Chr. 5 - 145587325 - 145835369 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

KCTD16
Genomic Map