Assay Details
| Gene Symbol: OSER1-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BC036500.1 | - | 3 - 4 | 475 | 98 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | oxidative stress responsive serine rich 1 | OSER1 |
C20orf111, HSPC207, Osr1, Perit1, dJ1183I21.1 |
-6.5kb | Anti-sense | Chr. 20 - 44195939 - 44217229 on NCBI Build GRCh38 |
| Downstream | ganglioside induced differentiation associated protein 1 like 1 | GDAP1L1 |
dJ881L22.1, dJ995J12.1.1 |
23.4kb | Sense | Chr. 20 - 44247099 - 44280917 on NCBI Build GRCh38 |
More Information
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC2000007447.hg.1 | ENST00000435163.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007447.hg.1 | ENST00000435163.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007447.hg.1 | ENST00000435163.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007447.hg.1 | ENST00000435163.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007447.hg.1 | ENST00000435163.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007447.hg.1 | ENST00000435163.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007447.hg.1 | ENST00000435163.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007447.hg.1 | ENST00000435163.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007447.hg.1 | ENST00000435163.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007447.hg.1 | ENST00000435163.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007447.hg.1 | ENST00000435163.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007447.hg.1 | ENST00000435163.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007447.hg.1 | ENST00000437730.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007447.hg.1 | ENST00000437730.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007447.hg.1 | ENST00000437730.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007447.hg.1 | ENST00000437730.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007447.hg.1 | ENST00000437730.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007447.hg.1 | ENST00000437730.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007447.hg.1 | ENST00000437730.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007447.hg.1 | ENST00000437730.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007447.hg.1 | ENST00000437730.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007447.hg.1 | ENST00000437730.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007447.hg.1 | ENST00000437730.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007447.hg.1 | ENST00000437730.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007447.hg.1 | ENST00000437730.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007447.hg.1 | ENST00000437730.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007447.hg.1 | ENST00000439943.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007447.hg.1 | ENST00000439943.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007447.hg.1 | ENST00000439943.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007447.hg.1 | ENST00000439943.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007447.hg.1 | ENST00000439943.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007447.hg.1 | ENST00000439943.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007447.hg.1 | ENST00000439943.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007447.hg.1 | ENST00000439943.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007447.hg.1 | ENST00000439943.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007447.hg.1 | ENST00000439943.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007447.hg.1 | ENST00000439943.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007447.hg.1 | ENST00000439943.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007447.hg.1 | ENST00000439943.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007447.hg.1 | ENST00000439943.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007447.hg.1 | ENST00000442383.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007447.hg.1 | ENST00000442383.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007447.hg.1 | ENST00000442383.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007447.hg.1 | ENST00000442383.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007447.hg.1 | ENST00000442383.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007447.hg.1 | ENST00000442383.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007447.hg.1 | ENST00000442383.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007447.hg.1 | ENST00000623343.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007447.hg.1 | ENST00000623343.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007447.hg.1 | ENST00000623343.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007447.hg.1 | ENST00000623343.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007447.hg.1 | ENST00000623343.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007447.hg.1 | ENST00000623343.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007447.hg.1 | ENST00000623343.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007447.hg.1 | NR_038337.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007447.hg.1 | NR_038337.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007447.hg.1 | NR_038337.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007447.hg.1 | NR_038337.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007447.hg.1 | NR_038337.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007447.hg.1 | NR_038337.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007447.hg.1 | NR_038337.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007447.hg.1 | NR_038337.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007447.hg.1 | NR_038337.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007447.hg.1 | NR_038338.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007447.hg.1 | NR_038338.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007447.hg.1 | NR_038338.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007447.hg.1 | NR_038338.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007447.hg.1 | NR_038338.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007447.hg.1 | NR_038338.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007447.hg.1 | NR_038338.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007447.hg.1 | NR_038338.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007447.hg.1 | NR_038339.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007447.hg.1 | NR_038339.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007447.hg.1 | NR_038339.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007447.hg.1 | NR_038339.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007447.hg.1 | NR_038339.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007447.hg.1 | NR_038339.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007447.hg.1 | NR_038339.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007447.hg.1 | ENST00000435163.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007447.hg.1 | ENST00000435163.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007447.hg.1 | ENST00000435163.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007447.hg.1 | ENST00000435163.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007447.hg.1 | ENST00000435163.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007447.hg.1 | ENST00000435163.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007447.hg.1 | ENST00000435163.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007447.hg.1 | ENST00000435163.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007447.hg.1 | ENST00000435163.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007447.hg.1 | ENST00000435163.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007447.hg.1 | ENST00000435163.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007447.hg.1 | ENST00000435163.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007447.hg.1 | ENST00000437730.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007447.hg.1 | ENST00000437730.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007447.hg.1 | ENST00000437730.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007447.hg.1 | ENST00000437730.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007447.hg.1 | ENST00000437730.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007447.hg.1 | ENST00000437730.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007447.hg.1 | ENST00000437730.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007447.hg.1 | ENST00000437730.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007447.hg.1 | ENST00000437730.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007447.hg.1 | ENST00000437730.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007447.hg.1 | ENST00000437730.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007447.hg.1 | ENST00000437730.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007447.hg.1 | ENST00000437730.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007447.hg.1 | ENST00000437730.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007447.hg.1 | ENST00000439943.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007447.hg.1 | ENST00000439943.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007447.hg.1 | ENST00000439943.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007447.hg.1 | ENST00000439943.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007447.hg.1 | ENST00000439943.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007447.hg.1 | ENST00000439943.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007447.hg.1 | ENST00000439943.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007447.hg.1 | ENST00000439943.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007447.hg.1 | ENST00000439943.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007447.hg.1 | ENST00000439943.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007447.hg.1 | ENST00000439943.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007447.hg.1 | ENST00000439943.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007447.hg.1 | ENST00000439943.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007447.hg.1 | ENST00000439943.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007447.hg.1 | ENST00000442383.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007447.hg.1 | ENST00000442383.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007447.hg.1 | ENST00000442383.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007447.hg.1 | ENST00000442383.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007447.hg.1 | ENST00000442383.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007447.hg.1 | ENST00000442383.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007447.hg.1 | ENST00000442383.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007447.hg.1 | ENST00000623343.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007447.hg.1 | ENST00000623343.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007447.hg.1 | ENST00000623343.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007447.hg.1 | ENST00000623343.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007447.hg.1 | ENST00000623343.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007447.hg.1 | ENST00000623343.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007447.hg.1 | ENST00000623343.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007447.hg.1 | NR_038337.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007447.hg.1 | NR_038337.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007447.hg.1 | NR_038337.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007447.hg.1 | NR_038337.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007447.hg.1 | NR_038337.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007447.hg.1 | NR_038337.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007447.hg.1 | NR_038337.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007447.hg.1 | NR_038337.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007447.hg.1 | NR_038337.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007447.hg.1 | NR_038338.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007447.hg.1 | NR_038338.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007447.hg.1 | NR_038338.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007447.hg.1 | NR_038338.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007447.hg.1 | NR_038338.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007447.hg.1 | NR_038338.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007447.hg.1 | NR_038338.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007447.hg.1 | NR_038338.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007447.hg.1 | NR_038339.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007447.hg.1 | NR_038339.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007447.hg.1 | NR_038339.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007447.hg.1 | NR_038339.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007447.hg.1 | NR_038339.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007447.hg.1 | NR_038339.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007447.hg.1 | NR_038339.2 | Refseq |

OSER1-AS1
Genomic Map