Assay Details
| Gene Symbol: USP39 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK057698.1 | - | 1 | 2049 | 66 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | chromosome 2 open reading frame 68 | C2orf68 | HCRCN81 | -34.3kb | Anti-sense | Chr. 2 - 85605253 - 85612073 on NCBI Build GRCh38 |
| Downstream | surfactant protein B | SFTPB |
PSP-B, SFTB3, SFTP3, SMDP1, SP-B |
8.0kb | Anti-sense | Chr. 2 - 85657317 - 85668741 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

USP39
Genomic Map