Assay Details
| Gene Symbol: RUNX1T1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK057707.1 | - | 1 | 2247 | 103 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | solute carrier family 26 member 7 | SLC26A7 | SUT2 | -694.7kb | Anti-sense | Chr. 8 - 91209494 - 91398155 on NCBI Build GRCh38 |
| Downstream | uncharacterized LOC102724710 | LOC102724710 | 470.2kb | Sense | Chr. 8 - 92565443 - 92655494 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

RUNX1T1
Genomic Map