Assay Details
| Gene Symbol: SEC14L2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK092576.1 | - | 1 | 550 | 90 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | ring finger protein 215 | RNF215 | -34.3kb | Anti-sense | Chr. 22 - 30378812 - 30387313 on NCBI Build GRCh38 | |
| Downstream | mitochondrial fission process 1 | MTFP1 |
HSPC242, MTP18 |
1.8kb | Sense | Chr. 22 - 30425623 - 30429054 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

SEC14L2
Genomic Map