Assay Details
| Gene Symbol: LIMK2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK092838.1 | - | 1 | 447 | 78 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | ring finger protein 185 | RNF185 | -70.8kb | Sense | Chr. 22 - 31160152 - 31207019 on NCBI Build GRCh38 | |
| Downstream | phosphoinositide-3-kinase interacting protein 1 | PIK3IP1 |
HGFL, hHGFL(S) |
1.5kb | Anti-sense | Chr. 22 - 31281593 - 31292534 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

LIMK2
Genomic Map