Assay Details
Gene Symbol: KCNJ15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK092958.1 | - | 1 | 2056 | 83 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | Down syndrome critical region 10 (non-protein coding) | DSCR10 | -62.1kb | Sense | Chr. 21 - 38206156 - 38208644 on NCBI Build GRCh38 | |
Downstream | long intergenic non-protein coding RNA 1423 | LINC01423 | 53.3kb | Anti-sense | Chr. 21 - 38326359 - 38346076 on NCBI Build GRCh38 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() |
