Assay Details
Gene Symbol: CCDC155 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK093322.1 | - | 2 - 3 | 804 | 86 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | uncharacterized LOC101928295 | LOC101928295 | -29.0kb | Anti-sense | Chr. 19 - 49368705 - 49388081 on NCBI Build GRCh38 | |
Downstream | golgi associated olfactory signaling regulator | GFY | Goofy | 3.3kb | Sense | Chr. 19 - 49425399 - 49429064 on NCBI Build GRCh38 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
