Assay Details
Gene Symbol: LOC100289347 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK094268.1 | - | 1 | 1753 | 100 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | cyclin H | CCNH |
CAK, CycH, p34, p37 |
-726.1kb | Sense | Chr. 5 - 87311480 - 87413033 on NCBI Build GRCh38 |
Downstream | transmembrane protein 161B | TMEM161B |
FLB3342, PRO1313 |
44.3kb | Sense | Chr. 5 - 88189419 - 88268917 on NCBI Build GRCh38 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
MicroArray Probes:
Agilent
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Human Whole Genome ChIP Tiling Array | A_17_P15718229 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P04081132 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P15718234 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P24261434 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P15718231 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P24261363 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P04081123 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P24261417 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P24261436 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P24261446 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P24261370 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P24261358 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P24261398 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P04081149 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P24261433 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P04081131 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P24261389 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P24261431 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P24261343 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P04081102 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P24261352 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P04081138 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P24261432 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P24261437 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P04081113 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P24261362 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P04081146 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P04081207 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P24261407 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P04081119 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P04081155 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P04081120 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P04081108 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P24261366 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P04081174 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P04081183 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P04081173 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P24261439 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P24261404 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P24261373 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P24261351 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P04081166 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P04081143 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P04081093 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P04081133 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P24261344 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P24261457 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P04081148 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P04081089 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P04081176 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P24261442 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P04081180 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P24261462 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P04081109 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P24261381 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P24261422 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P24261438 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P24261385 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P04081163 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P04081179 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P04081134 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P04081128 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P04081160 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P04081129 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P04081181 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P04081175 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P24261448 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P24261444 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P24261452 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P04081157 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P24261453 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P24261427 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P24261367 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P24261440 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P24261428 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P24261423 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P24261415 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P24261430 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P24261369 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P04081092 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P24261461 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P04081130 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P04081090 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P04081153 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P24261456 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P24261463 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P04081107 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P24261397 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P24261350 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P24261435 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P24261413 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P04081097 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P24261361 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P04081091 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P24261449 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P24261386 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P24261429 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P24261387 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P24261348 | - | - |
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 600073 | - | - |
