Assay Details
| Gene Symbol: XPNPEP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK094900.1 | - | 1 | 1555 | 99 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | long intergenic non-protein coding RNA 1435 | LINC01435 | TCONS_00018040 | -1.8mb | Sense | Chr. 10 - 107871576 - 108069293 on NCBI Build GRCh38 |
| Downstream | ADD3 antisense RNA 1 | ADD3-AS1 | 78.7kb | Sense | Chr. 10 - 109945559 - 110008381 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

XPNPEP1
Genomic Map