Assay Details
Gene Symbol: KCNK9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK095204.1 | - | 1 | 1542 | 75 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | collagen type XXII alpha 1 | COL22A1 | -696.4kb | Sense | Chr. 8 - 138588235 - 138914087 on NCBI Build GRCh38 | |
Downstream | trafficking protein particle complex 9 | TRAPPC9 |
IBP, IKBKBBP, MRT13, NIBP, T1, TRS120 |
116.0kb | Sense | Chr. 8 - 139727725 - 140458579 on NCBI Build GRCh38 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() |
