Assay Details
| Gene Symbol: MID1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AY660579.1 | - | 2 - 3 | 326 | 99 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | chloride voltage-gated channel 4 | CLCN4 |
CLC4, ClC-4, ClC-4A |
-622.4kb | Anti-sense | Chr. X - 10156945 - 10237660 on NCBI Build GRCh38 |
| Downstream | holocytochrome c synthase | HCCS |
CCHL, LSDMCA1, MCOPS7, MLS |
4.3kb | Anti-sense | Chr. X - 11111286 - 11123086 on NCBI Build GRCh38 |
More Information
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009064.hg.1 | ENST00000433747.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009064.hg.1 | ENST00000433747.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009064.hg.1 | ENST00000433747.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009064.hg.1 | ENST00000433747.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009064.hg.1 | ENST00000433747.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009064.hg.1 | ENST00000433747.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009064.hg.1 | ENST00000433747.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009064.hg.1 | ENST00000433747.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009064.hg.1 | ENST00000433747.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009064.hg.1 | ENST00000433747.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009064.hg.1 | ENST00000433747.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009064.hg.1 | ENST00000433747.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009064.hg.1 | ENST00000608176.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009064.hg.1 | ENST00000608176.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009064.hg.1 | ENST00000608176.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009064.hg.1 | ENST00000608176.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009064.hg.1 | ENST00000608176.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009064.hg.1 | ENST00000608176.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009064.hg.1 | ENST00000608176.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009064.hg.1 | ENST00000608176.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009064.hg.1 | ENST00000608176.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009064.hg.1 | ENST00000608176.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009064.hg.1 | ENST00000608176.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009064.hg.1 | ENST00000608176.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009064.hg.1 | ENST00000608176.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009064.hg.1 | ENST00000608176.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009064.hg.1 | ENST00000608576.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009064.hg.1 | ENST00000608576.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009064.hg.1 | ENST00000608576.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009064.hg.1 | ENST00000608576.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009064.hg.1 | ENST00000608576.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009064.hg.1 | ENST00000608576.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009064.hg.1 | ENST00000608576.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009064.hg.1 | ENST00000608576.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009064.hg.1 | ENST00000608576.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009064.hg.1 | ENST00000608576.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009064.hg.1 | ENST00000608576.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009064.hg.1 | ENST00000608576.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009064.hg.1 | ENST00000608576.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009064.hg.1 | ENST00000608576.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009064.hg.1 | ENST00000608576.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009064.hg.1 | ENST00000608576.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009064.hg.1 | ENST00000608916.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009064.hg.1 | ENST00000608916.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009064.hg.1 | ENST00000608916.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009064.hg.1 | ENST00000608916.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009064.hg.1 | ENST00000608916.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009064.hg.1 | ENST00000608916.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009064.hg.1 | ENST00000608916.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009064.hg.1 | ENST00000433747.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009064.hg.1 | ENST00000433747.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009064.hg.1 | ENST00000433747.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009064.hg.1 | ENST00000433747.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009064.hg.1 | ENST00000433747.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009064.hg.1 | ENST00000433747.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009064.hg.1 | ENST00000433747.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009064.hg.1 | ENST00000433747.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009064.hg.1 | ENST00000433747.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009064.hg.1 | ENST00000433747.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009064.hg.1 | ENST00000433747.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009064.hg.1 | ENST00000433747.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009064.hg.1 | ENST00000608176.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009064.hg.1 | ENST00000608176.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009064.hg.1 | ENST00000608176.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009064.hg.1 | ENST00000608176.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009064.hg.1 | ENST00000608176.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009064.hg.1 | ENST00000608176.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009064.hg.1 | ENST00000608176.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009064.hg.1 | ENST00000608176.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009064.hg.1 | ENST00000608176.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009064.hg.1 | ENST00000608176.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009064.hg.1 | ENST00000608176.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009064.hg.1 | ENST00000608176.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009064.hg.1 | ENST00000608176.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009064.hg.1 | ENST00000608176.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009064.hg.1 | ENST00000608576.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009064.hg.1 | ENST00000608576.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009064.hg.1 | ENST00000608576.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009064.hg.1 | ENST00000608576.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009064.hg.1 | ENST00000608576.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009064.hg.1 | ENST00000608576.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009064.hg.1 | ENST00000608576.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009064.hg.1 | ENST00000608576.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009064.hg.1 | ENST00000608576.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009064.hg.1 | ENST00000608576.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009064.hg.1 | ENST00000608576.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009064.hg.1 | ENST00000608576.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009064.hg.1 | ENST00000608576.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009064.hg.1 | ENST00000608576.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009064.hg.1 | ENST00000608576.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009064.hg.1 | ENST00000608576.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009064.hg.1 | ENST00000608916.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009064.hg.1 | ENST00000608916.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009064.hg.1 | ENST00000608916.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009064.hg.1 | ENST00000608916.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009064.hg.1 | ENST00000608916.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009064.hg.1 | ENST00000608916.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009064.hg.1 | ENST00000608916.1 | Ensembl |

MID1
Genomic Map