Assay Details
| Gene Symbol: TTC3P1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_030737.1 | 1 | 2205 | 57 | ||
| GenBank mRNA | AK304575.1 | - | 1 | 1194 | 57 | |
| AY672106.1 | - | 1 | 51 | 57 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | BRAF pseudogene 1 | BRAFP1 | BRAF2, BRAFPS2 | -154.5kb | Anti-sense | Chr. X - 75582654 - 75586009 on NCBI Build GRCh38 | 
| Downstream | MAGE family member E2 | MAGEE2 | HCA3 | 42.1kb | Sense | Chr. X - 75782988 - 75785244 on NCBI Build GRCh38 | 
 Back To Top
 Back To TopMore Information
Updated Mapping Information
 This assay targets the same location as Clariom D transcript cluster(s) TC0X00010112.hg.1
					            	This assay targets the same location as Clariom D transcript cluster(s) TC0X00010112.hg.1This assay targets the same location as HTA transcript cluster(s) TC0X001166.hg.1
| Set Membership: |  Amplicon length less than or equal to 70  3' Most  Detect Genomic DNA  Homo sapiens  Both primers and probe map within a single exon | 
MicroArray Probes:
					
						
							Agilent
					
						
							Affymetrix
					
						
							ArrayStar
					
						
							Illumina
					
					
					
						
							
								
						
					
						
							
								
						
					
						
							
								
						
					
						
							
								
						
					
				
			| Array Name | Probe IDs | Targets | Target Type | 
|---|---|---|---|
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P17359320 | - | - | 
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P11793687 | - | - | 
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P17359283 | - | - | 
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P11793672 | - | - | 
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P11793697 | - | - | 
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P11793729 | - | - | 
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P32510720 | - | - | 
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P32510632 | - | - | 
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P11793649 | - | - | 
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P11793671 | - | - | 
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P17359325 | - | - | 
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P11793702 | - | - | 
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P11793717 | - | - | 
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P11793658 | - | - | 
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P32510659 | - | - | 
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P32510668 | - | - | 
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P11793700 | - | - | 
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P32510628 | - | - | 
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P32510626 | - | - | 
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P11793677 | - | - | 
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P11793667 | - | - | 
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P11793705 | - | - | 
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P11793726 | - | - | 
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P11793647 | - | - | 
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P11793655 | - | - | 
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P32510713 | - | - | 
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P11793690 | - | - | 
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P32510619 | - | - | 
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P32510688 | - | - | 
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P32510714 | - | - | 
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P11793692 | - | - | 
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P32510620 | - | - | 
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P32510697 | - | - | 
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P32510725 | - | - | 
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P11793663 | - | - | 
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P11793652 | - | - | 
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P32510698 | - | - | 
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P11793661 | - | - | 
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P11793693 | - | - | 
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P32510710 | - | - | 
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P32510673 | - | - | 
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P17359294 | - | - | 
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P32510724 | - | - | 
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P11793679 | - | - | 
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P17359321 | - | - | 
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P17359287 | - | - | 
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P11793669 | - | - | 
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P32510695 | - | - | 
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P32510649 | - | - | 
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P11793653 | - | - | 
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P17359291 | - | - | 
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P11793659 | - | - | 
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P32510677 | - | - | 
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P11793651 | - | - | 
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P11793721 | - | - | 
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P11793699 | - | - | 
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P11793715 | - | - | 
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P32510621 | - | - | 
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P11793695 | - | - | 
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P11793727 | - | - | 
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P32510716 | - | - | 
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P11793723 | - | - | 
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P32510705 | - | - | 
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P32510651 | - | - | 
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P32510717 | - | - | 
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P11793704 | - | - | 
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P11793686 | - | - | 
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P32510622 | - | - | 
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P11793685 | - | - | 
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P11793646 | - | - | 
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P11793696 | - | - | 
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P11793673 | - | - | 
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P11793678 | - | - | 
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P11793694 | - | - | 
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P32510701 | - | - | 
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P11793716 | - | - | 
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P32510704 | - | - | 
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P11793701 | - | - | 
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P11793710 | - | - | 
| Agilent Whole Human Genome Oligo Microarray 4x44K | 41528 | - | - | 
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32510673 | - | - | 
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11793702 | - | - | 
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11793671 | - | - | 
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32510713 | - | - | 
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32510720 | - | - | 
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11793705 | - | - | 
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32510716 | - | - | 
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32510714 | - | - | 
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11793697 | - | - | 
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32510668 | - | - | 
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32510701 | - | - | 
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11793677 | - | - | 
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11793678 | - | - | 
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11793730 | - | - | 
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32510704 | - | - | 
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P17359320 | - | - | 
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11793721 | - | - | 
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11793679 | - | - | 
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32510724 | - | - | 
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11793715 | - | - | 
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11793690 | - | - | 
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32510659 | - | - | 
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11793716 | - | - | 
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32510698 | - | - | 
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11793710 | - | - | 
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11793692 | - | - | 
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11793727 | - | - | 
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32510688 | - | - | 
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11793731 | - | - | 
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11793685 | - | - | 
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P17359325 | - | - | 
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32510726 | - | - | 
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11793701 | - | - | 
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11793673 | - | - | 
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P17359321 | - | - | 
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32510710 | - | - | 
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32510677 | - | - | 
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11793693 | - | - | 
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11793687 | - | - | 
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11793723 | - | - | 
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11793708 | - | - | 
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11793729 | - | - | 
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11793694 | - | - | 
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11793682 | - | - | 
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11793696 | - | - | 
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11793695 | - | - | 
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32510705 | - | - | 
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11793726 | - | - | 
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32510695 | - | - | 
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11793683 | - | - | 
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11793672 | - | - | 
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11793676 | - | - | 
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11793669 | - | - | 
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32510717 | - | - | 
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11793733 | - | - | 
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11793700 | - | - | 
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11793717 | - | - | 
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32510651 | - | - | 
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32510726 | - | - | 
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P17359325 | - | - | 
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11793676 | - | - | 
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P17359320 | - | - | 
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32510695 | - | - | 
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11793690 | - | - | 
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11793730 | - | - | 
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32510713 | - | - | 
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11793673 | - | - | 
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32510720 | - | - | 
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11793701 | - | - | 
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32510688 | - | - | 
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32510673 | - | - | 
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32510668 | - | - | 
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11793716 | - | - | 
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11793697 | - | - | 
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11793731 | - | - | 
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32510714 | - | - | 
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11793683 | - | - | 
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11793687 | - | - | 
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11793679 | - | - | 
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32510659 | - | - | 
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11793693 | - | - | 
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11793671 | - | - | 
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11793729 | - | - | 
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32510724 | - | - | 
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32510677 | - | - | 
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11793682 | - | - | 
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32510704 | - | - | 
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11793702 | - | - | 
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32510705 | - | - | 
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32510710 | - | - | 
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32510717 | - | - | 
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11793677 | - | - | 
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32510698 | - | - | 
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11793721 | - | - | 
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32510716 | - | - | 
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11793717 | - | - | 
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11793669 | - | - | 
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32510651 | - | - | 
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11793726 | - | - | 
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11793695 | - | - | 
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11793715 | - | - | 
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11793710 | - | - | 
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11793692 | - | - | 
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11793727 | - | - | 
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11793733 | - | - | 
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11793685 | - | - | 
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11793696 | - | - | 
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11793678 | - | - | 
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11793672 | - | - | 
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11793723 | - | - | 
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32510701 | - | - | 
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11793705 | - | - | 
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11793694 | - | - | 
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11793700 | - | - | 
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11793708 | - | - | 
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P17359321 | - | - | 
| Whole Human Genome Oligo Microarray G4112A | A_32_P14201 | - | - | 
| Whole Human Genome Oligo Microarray G4112A | A_32_P83453 | - | - | 
| Array Name | Probe IDs | Targets | Target Type | 
|---|---|---|---|
| Affymetrix Human Genome U219 Array | 11760965_at | - | - | 
| Affymetrix Human Genome U219 Array | 11760965_PM_at | - | - | 
| Affymetrix Human Transcriptome Array 2.0 | PSR0X015738.hg.1 | TC0X001166.hg.1 | HTA TC | 
| Affymetrix Human Transcriptome Array 2.0 | PSR0X015738.hg.1 | NR_030737.1 | Refseq | 
| Affymetrix Human Clariom D Assay | PSR0X00169006.hg.1 | NR_030737.1 | Refseq | 
| Affymetrix Human Clariom D Assay | PSR0X00169006.hg.1 | NR_030737.1 | Refseq | 
| Affymetrix Human Clariom D Assay | PSR0X00169006.hg.1 | TC0X00010112.hg.1 | Clariom D TC | 
| Affymetrix Human Clariom D Assay | PSR0X00169007.hg.1 | ENST00000399586.2 | Ensembl | 
| Affymetrix Human Clariom D Assay | PSR0X00169007.hg.1 | NR_030737.1 | Refseq | 
| Affymetrix Human Clariom D Assay | PSR0X00169007.hg.1 | ENST00000399586.2 | Ensembl | 
| Affymetrix Human Clariom D Assay | PSR0X00169007.hg.1 | NR_030737.1 | Refseq | 
| Affymetrix Human Clariom D Assay | PSR0X00169007.hg.1 | TC0X00010112.hg.1 | Clariom D TC | 
| Array Name | Probe IDs | Targets | Target Type | 
|---|---|---|---|
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A040095 | - | - | 
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A040095 | - | - | 
| Array Name | Probe IDs | Targets | Target Type | 
|---|---|---|---|
| HumanHT-12 v4 Expression BeadChip | ILMN_3310271 | AY672106.1 | human_transcriptome | 
| HumanHT-12 v4 Expression BeadChip | ILMN_3310271 | NR_030737.1 | human_transcriptome | 
| HumanHT-12 v4 Expression BeadChip | ILMN_3310271 | AK304575.1 | human_transcriptome | 
| HumanHT-12 V4.0 expression beadchip | ILMN_3310271 | AY672106.1 | human_transcriptome | 
| HumanHT-12 V4.0 expression beadchip | ILMN_3310271 | NR_030737.1 | human_transcriptome | 
| HumanHT-12 V4.0 expression beadchip | ILMN_3310271 | AK304575.1 | human_transcriptome | 
| Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_3310271 | AY672106.1 | human_transcriptome | 
| Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_3310271 | NR_030737.1 | human_transcriptome | 
| Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_3310271 | AK304575.1 | human_transcriptome | 
| Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_3310271 | AY672106.1 | human_transcriptome | 
| Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_3310271 | NR_030737.1 | human_transcriptome | 
| Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_3310271 | AK304575.1 | human_transcriptome | 
 Back To Top
 Back To Top
 TTC3P1
TTC3P1 Genomic Map
Genomic Map Back To Top
 Back To Top