Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010589.hg.1 |
ENST00000527828.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010589.hg.1 |
ENST00000527828.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010589.hg.1 |
ENST00000527828.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010589.hg.1 |
ENST00000527828.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010589.hg.1 |
ENST00000527828.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010589.hg.1 |
ENST00000527828.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010589.hg.1 |
ENST00000527828.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010589.hg.1 |
ENST00000527828.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010589.hg.1 |
ENST00000534059.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010589.hg.1 |
ENST00000534059.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010589.hg.1 |
ENST00000534059.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010589.hg.1 |
ENST00000534059.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010589.hg.1 |
ENST00000534059.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010589.hg.1 |
ENST00000534059.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010589.hg.1 |
ENST00000527828.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010589.hg.1 |
ENST00000527828.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010589.hg.1 |
ENST00000527828.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010589.hg.1 |
ENST00000527828.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010589.hg.1 |
ENST00000527828.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010589.hg.1 |
ENST00000527828.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010589.hg.1 |
ENST00000527828.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010589.hg.1 |
ENST00000527828.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010589.hg.1 |
ENST00000534059.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010589.hg.1 |
ENST00000534059.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010589.hg.1 |
ENST00000534059.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010589.hg.1 |
ENST00000534059.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010589.hg.1 |
ENST00000534059.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010589.hg.1 |
ENST00000534059.1 |
Ensembl |